Merck
  • Therapeutic potential of selenium and tellurium compounds: opportunities yet unrealised.
Therapeutic potential of selenium and tellurium compounds: opportunities yet unrealised.
Dalton transactions (Cambridge, England : 2003) (2012-01-19)
Edward R T Tiekink
ABSTRACT
Despite being disparaged for their malodorous and toxic demeanour, compounds of selenium, a bio-essential element, and tellurium, offer possibilities as therapeutic agents. Herein, their potential use as drugs, for example, as anti-viral, anti-microbial, anti-inflammatory agents, etc., will be surveyed along with a summary of the established biological functions of selenium. The natural biological functions of tellurium remain to be discovered.
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis