Merck
Nitric oxide signaling in aluminum stress in plants.
Protoplasma (2011-08-19)
Huyi He, Jie Zhan, Longfei He, Minghua Gu
ABSTRACT
Nitric oxide (NO) is a ubiquitous signal molecule involved in multiple plant responses to environmental stress. In the recent years, the regulating role of NO on heavy metal toxicity in plants is realized increasingly, but knowledge of NO in alleviating aluminum (Al) toxicity is quite limited. In this article, NO homeostasis between its biosynthesis and elimination in plants is presented. Some genes involved in NO/Al network and their expressions are also introduced. Furthermore, the role of NO in Al toxicity and the functions in Al tolerance are discussed. It is proposed that Al toxicity may disrupt NO homeostasis, leading to endogenous NO concentration being lower than required for root elongation in plants. There are many evidences that pointed out that the exogenous NO treatments improve Al tolerance in plants through activating antioxidative capacity to eliminate reactive oxygen species. Most of the work with respect to NO regulating pathways and functions still has to be done in the future.
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, wire reel, 5m, diameter 2.0mm, as drawn, 99.999%
Aluminum, wire reel, 5m, diameter 3.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, microleaf disks, 10mm, thinness 1.0μm, specific density 269.9μg/cm2, removable support, 99.999%
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, rod, 500mm, diameter 5.0mm, as drawn, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, rod, 100mm, diameter 12.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.05mm, as drawn, 99.99+%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, sphere, 100pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 200mm, diameter 20.0mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 28mm, as drawn, 99.999%