Merck
Size-dependent in vivo toxicity of PEG-coated gold nanoparticles.
International journal of nanomedicine (2011-10-07)
Xiao-Dong Zhang, Di Wu, Xiu Shen, Pei-Xun Liu, Na Yang, Bin Zhao, Hao Zhang, Yuan-Ming Sun, Liang-An Zhang, Fei-Yue Fan
ABSTRACT
Gold nanoparticle toxicity research is currently leading towards the in vivo experiment. Most toxicology data show that the surface chemistry and physical dimensions of gold nanoparticles play an important role in toxicity. Here, we present the in vivo toxicity of 5, 10, 30, and 60 nm PEG-coated gold nanoparticles in mice. Animal survival, weight, hematology, morphology, organ index, and biochemistry were characterized at a concentration of 4000 μg/kg over 28 days. The PEG-coated gold particles did not cause an obvious decrease in body weight or appreciable toxicity even after their breakdown in vivo. Biodistribution results show that 5 nm and 10 nm particles accumulated in the liver and that 30 nm particles accumulated in the spleen, while the 60 nm particles did not accumulate to an appreciable extent in either organ. Transmission electron microscopic observations showed that the 5, 10, 30, and 60 nm particles located in the blood and bone marrow cells, and that the 5 and 60 nm particles aggregated preferentially in the blood cells. The increase in spleen index and thymus index shows that the immune system can be affected by these small nanoparticles. The 10 nm gold particles induced an increase in white blood cells, while the 5 nm and 30 nm particles induced a decrease in white blood cells and red blood cells. The biochemistry results show that the 10 nm and 60 nm PEG-coated gold nanoparticles caused a significant increase in alanine transaminase and aspartate transaminase levels, indicating slight damage to the liver. The toxicity of PEG-coated gold particles is complex, and it cannot be concluded that the smaller particles have greater toxicity. The toxicity of the 10 nm and 60 nm particles was obviously higher than that of the 5 nm and 30 nm particles. The metabolism of these particles and protection of the liver will be more important issues for medical applications of gold-based nanomaterials in future.
Sigma-Aldrich
Gold nanoparticles, 10 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, carboxylic acid functionalized, PEG 3000 coated, OD 50, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, OD 50, methyl terminated, PEG 5000 coated, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, amine functionalized, PEG 5000 coated, OD 50, dispersion in H2O
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, amine functionalized, PEG 3000 coated, OD 50, dispersion in H2O