Merck
  • Effect of silver nanoparticles on rice (Oryza sativa L. cv. KDML 105) seed germination and seedling growth.
Effect of silver nanoparticles on rice (Oryza sativa L. cv. KDML 105) seed germination and seedling growth.
Ecotoxicology and environmental safety (2014-04-15)
Pakvirun Thuesombat, Supot Hannongbua, Sanong Akasit, Supachitra Chadchawan
ABSTRACT
With the advances in nanotechnology, silver nanoparticles (AgNPs) have been applied in many industries, increasing their potential exposure level in the environment, yet their environmental safety remains poorly evaluated. The possible effects of different sized AgNPs (20, 30-60, 70-120 and 150nm diameter) on jasmine rice, Oryza sativa L. cv. KDML 105, were investigated at different concentrations (0.1, 1, 10, 100 and 1000mg/L) upon seed germination and seedling growth. The results revealed that the level of seed germination and subsequent growth of those seedlings that germinated were both decreased with increasing sizes and concentrations of AgNPs. Based on the analysis of AgNPs accumulation in plant tissues, it implied that the higher uptake was found when the seeds were treated with the smaller AgNPs, 20nm diameter AgNPs, but it was trapped in the roots rather than transported to the leaves. These resulted in the less negative effects on seedling growth, when compared to the seed soaking with the larger AgNPs with 150nm diameter. The negative effects of AgNPs were supported by leaf cell deformation when rice seeds were treated with 150-nm-diameter AgNP at the concentration of 10 or 100mg/L during seed germination. These results further strengthen our understanding of environmental safety information with respect to nanomaterials.
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, conductive paste
Silver, tube, 100mm, outside diameter 0.7mm, inside diameter 0.3mm, wall thickness 0.2mm, as drawn, 99.95+%
Silver, foil, 8mm disks, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%
Silver, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.008mm, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.015mm, 99.97%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silver, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%
Silver, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, not light tested, 25x25mm, thickness 0.006mm, 99.97%
Silver, foil, not light tested, 150x150mm, thickness 0.02mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%