Merck
  • A rapid and sensitive method for the detection of Xanthomonas fragariae, causal agent of angular leafspot disease in strawberry plants.
A rapid and sensitive method for the detection of Xanthomonas fragariae, causal agent of angular leafspot disease in strawberry plants.
Journal of microbiological methods (2004-07-06)
Anna Stöger, Werner Ruppitsch
ABSTRACT
Angular leaf spot is a bacterial disease caused by Xanthomonas fragariae. It has become a serious disease in the USA and Europe in recent years. Several detection procedures are described for this plant pathogen. However, they are either too time-consuming, too insensitive or impractical when handling a large number of samples routinely. Here we describe a modified protocol of the REDExtract-N-Amp Plant PCR-Kit for the detection of X. fragariae in planta and demonstrate that it provides greater sensitivity, speed and high throughput potential than methods previously described.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 500 amplifications