Merck
  • Superficial neuromasts facilitate non-visual feeding by larval striped bass (Morone saxatilis).
Superficial neuromasts facilitate non-visual feeding by larval striped bass (Morone saxatilis).
The Journal of experimental biology (2013-06-06)
Julia A Sampson, Jim Duston, Roger P Croll
ABSTRACT
To investigate whether mechanoreception is used in non-visual feeding in larval striped bass (Morone saxatilis), the ontogeny of superficial neuromasts along the lateral line was described using the vital stain FM1-43FX and fluorescent microscopy. The number of neuromasts visible along one flank increased from 11 at first feeding [5 to 7 days post-hatch (dph)] to >150 by the juvenile stage (27 dph). A neomycin dose response (0, 1, 2 and 5 mmol l(-1)) was evaluated for neuromast ablation of bass aged 10, 13, 17 and 20 dph. Using these same age groups, the ability of bass to catch Artemia salina prey in both dark and light tank-based feeding trials was compared between larvae with neuromasts ablated using neomycin (5 mmol l(-1)) and controls. Neomycin significantly reduced the incidence of feeding in the light and dark. Among larvae that fed, those in the dark treated with neomycin caught fewer Artemia (~5 prey h(-1); P<0.05) than controls (16 prey h(-1) at 10 dph; 72 prey h(-1) at 20 dph). In the light, by contrast, neomycin treatment had no significant effect on prey capture by larvae age 13 to 20 dph, but did inhibit feeding of 10 dph larvae. Verification that neomycin was specifically ablating the hair cells of superficial neuromasts and not affecting either neuromast innervation, olfactory pits, or taste cells was achieved by a combination of staining with FM1-43FX and immunocytochemistry for tubulin and the calcium binding proteins, S100 and calretinin.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard