Merck
  • Molecular diversity of Mycobacterium tuberculosis isolates from patients with tuberculosis in Honduras.
Molecular diversity of Mycobacterium tuberculosis isolates from patients with tuberculosis in Honduras.
BMC microbiology (2010-08-04)
Senia Rosales, Lelany Pineda-García, Solomon Ghebremichael, Nalin Rastogi, Sven E Hoffner
ABSTRACT
Tuberculosis persists as a public health problem in Honduras. A better knowledge of the molecular characteristics of Mycobacterium tuberculosis strains will contribute to understand the transmission dynamics of the disease within the country. The aim of this study was to provide an insight of the genetic biodiversity of M. tuberculosis clinical isolates collected in Honduras between 1994 and 2002. Genotyping was performed using spoligotyping and RFLP. The spoligotypes obtained were compared with the SITVIT2 proprietary database of the Pasteur Institute of Guadeloupe. Spoligotyping grouped 84% of the isolates into 27 clusters (2 to 43 strains per cluster). Of the 44 shared international types (SITs) identified among the Honduran stains, 8 SITs were newly identified either within the present study or after match with an orphan type previously identified in the SITVIT2 database. In addition, 16 patterns corresponded to orphan, previously unreported isolates.The Latin American Mediterranean (LAM) lineage was the most common in this study; 55% of the strains belonged to this family. Other genotypes found were Haarlem (16%), T (16%), X-clade (6%), Unknown signature (5%) and S (1%). Only one Beijing strain was identified (0.5%).We observed a high degree of diversity after characterizing the 43 isolates belonging to the main spoligotyping cluster (SIT 33, LAM3) with IS6110-RFLP. A total of 35 different RFLP-fingerprints were detected, of which 6 patterns corresponded to the same number of clusters comprising 14 strains. The findings obtained in this study show that tuberculosis transmission in Honduras is due to modern M. tuberculosis lineages with high level of biodiversity.
Product Description
ECL Detection Reagents, Cytiva RPN3004, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3005, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3001, pack of 1 ea
ECL Direct Nucleic Acid, Cytiva RPN3000, pack of 1 ea