Merck
  • A facile chemical approach for preparing a SERS active silver substrate.
A facile chemical approach for preparing a SERS active silver substrate.
Physical chemistry chemical physics : PCCP (2010-10-01)
Cheng Yang, Yu-Tao Xie, Matthew M F Yuen, Xiaomin Xiong, C P Wong
ABSTRACT
This communication describes a new surface-enhanced Raman scattering (SERS) active silver substrate prepared by iodination of the evaporated silver foil. After iodination, the morphology of the silver substrate undergoes a self-evolution process in which it displays accordingly the UV-vis absorption shift as well as the AFM topological test. Rhodamine 6G (R6G) is used as the probe molecule to evaluate the enhancement efficiency of the silver substrate at different self-evolution time intervals. The SERS intensity of R6G increases up to ∼29-fold and reaches a maximum after the substrate evolved for 24 h. This method is feasible for the production of an efficient SERS silver substrate.
Product Description
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, not light tested, 50x50mm, thickness 0.007mm, 99.97%
Silver, foil, not light tested, 50x50mm, thickness 0.002mm, 99.97%
Silver, foil, 10mm disks, thickness 0.02mm, 99.97%
Silver, foil, 0.5m coil, thickness 0.025mm, annealed, 99.95+%
Silver, foil, 10mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 50mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 25mm disks, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, 2m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 50mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 25mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 2m coil, thickness 0.0125mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.02mm, 99.97%
Silver, foil, 4mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.015mm, 99.97%
Silver, foil, 4mm disks, thickness 0.007mm, 99.97%
Silver, foil, 50mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 6mm disks, thickness 0.008mm, 99.97%
Silver, foil, 4mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 6mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 25mm disks, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 5m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 6mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.005mm, 99.97%
Silver, foil, 4mm disks, thickness 0.004mm, 99.97%