Merck
  • Self-limiting growth of metal fluoride thin films by oxidation reactions employing molecular precursors
Self-limiting growth of metal fluoride thin films by oxidation reactions employing molecular precursors
Physical review letters (2000-09-06)
Qiu, Lai, Yarmoff
ABSTRACT
FeF (2) films are grown by the reaction of XeF (2) and SeF (6) with iron foil. The growth initially follows the Mott-Cabrera parabolic rate law, indicating that the process is diffusion limited. At a certain film thickness, however, the growth abruptly stops, with the thickness using XeF (2) being nearly double that with SeF (6). It is suggested that the shutdown is due to the inability of the molecules to dissociate when too far from the substrate and that SeF (6) must approach more closely than XeF (2). This work suggests the use of molecular precursors to grow thin films via a self-limiting chemical process.
Product Description
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 15mm disks, thickness 0.1mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 25mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.025mm, as rolled, 99.99+%
Iron, foil, 50mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.20mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, light tested, 100x100mm, thickness 0.025mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, light tested, 100x100mm, thickness 0.038mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.003mm, 99.85%
Iron, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.85%
Iron, foil, light tested, 25x25mm, thickness 0.075mm, hard, 99.5%
Iron, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 100x100mm, thickness 0.125mm, hard, 99.5%
Iron, foil, not light tested, 50x50mm, thickness 0.003mm, 99.85%
Iron, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.008mm, 99.85%
Iron, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%