Merck
  • Tolrestat pharmacokinetic and pharmacodynamic effects on red blood cell sorbitol levels in normal volunteers and in patients with insulin-dependent diabetes.
Tolrestat pharmacokinetic and pharmacodynamic effects on red blood cell sorbitol levels in normal volunteers and in patients with insulin-dependent diabetes.
Clinical pharmacology and therapeutics (1995-12-01)
J M van Griensven, W J Jusko, H H Lemkes, R Kroon, C J Verhorst, S T Chiang, A F Cohen
ABSTRACT
To examine the effect of diabetes mellitus on the pharmacokinetics of tolrestat and to investigate its effect on red blood cell sorbitol levels according to a new pharmacodynamic model for this class of drugs. Single and multiple doses of tolrestat (200 mg/twice a day) were administered to 12 patients with insulin-dependent (type I) diabetes and 12 healthy volunteers in an open parallel trial. Tolrestat disposition was characterized by first-order absorption and biexponential disposition: In normal subjects the terminal disposition half-life (t1/2) was 13 +/- 3 hours (mean +/- SD) and the apparent oral clearance (CL/F) was 48 +/- 9 ml/hr/kg, similar to the values in patients with type I diabetes mellitus (t1/2 = 14 +/- 4 hours; CL/F = 55 +/- 10 ml/hr/kg). Red blood cell sorbitol concentrations, which declined because of tolrestat's inhibition of aldose reductase, were characterized by an indirect-response model including a 50% inhibition constant (IC50) for production of sorbitol by aldose reductase. The removal of sorbitol (kout) was slower in patients with diabetes. The plasma IC50 averaged 2.0 +/- 1.3 micrograms/ml in normal subjects and 2.5 +/- 1.9 micrograms/ml in patients with diabetes. IC50 values expressed in free (unbound) concentrations (fu = 0.64%), which ranged from 12 to 16 ng/ml, were similar to the in vitro IC50 for inhibition of sorbitol accumulation in human red blood cells. Tolrestat pharmacokinetics were similar in normal subjects and in patients with diabetes; however, the patients with diabetes had higher baseline sorbitol levels (11 versus 5 nmol/ml for normal subjects) and slower sorbitol efflux rates. The proposed biochemically realistic, dynamic model characterized well the red blood cell sorbitol response patterns after administration of single and multiple doses of tolrestat.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)