Merck
  • Fabrication of high-quality ZnTe nanowires toward high-performance rigid/flexible visible-light photodetectors.
Fabrication of high-quality ZnTe nanowires toward high-performance rigid/flexible visible-light photodetectors.
Optics express (2013-04-03)
Zhe Liu, Gui Chen, Bo Liang, Gang Yu, Hongtao Huang, Di Chen, Guozhen Shen
ABSTRACT
ZnTe is an important p-type semiconductor with great applications as field-effect transistors and photodetectors. In this paper, individual ZnTe nanowires based field-effect transistors was fabricated, showing evident p-type conductivity with an effect mobility of 11.3 cm(2)/Vs. Single ZnTe nanowire based photodetectors on rigid silicon substrate exhibited high sensitivity and excellent stability to visible incident light with responstivity and quantum efficiency as high as 1.87 × 10(5) A/W and 4.36 × 10(7)% respectively and are stable in a wide temperature range (25-250 °C). The polarization-sensitivity of the ZnTe nanowires was studied for the first time. The results revealed a periodic oscillation with the continuous variation of polarization angles. Besides, flexible photodetectors were also fabricated with the features of excellent flexibility, stability and sensitivity to visible incident light. Our work would enable application opportunities in using ZnTe nanowires for ultrahigh-performance photodetectors in scientific, commercial and industrial applications.
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis