Merck
  • Benzenesulfonamides: a unique class of chemokine receptor type 4 inhibitors.
Benzenesulfonamides: a unique class of chemokine receptor type 4 inhibitors.
ChemMedChem (2013-03-08)
Suazette Reid Mooring, Jin Liu, Zhongxing Liang, Jeffrey Ahn, Samuel Hong, Younghyoun Yoon, James P Snyder, Hyunsuk Shim
ABSTRACT
The interaction of CXCR4 with CXCL12 (SDF-1) plays a critical role in cancer metastasis by facilitating the homing of tumor cells to metastatic sites. Based on our previously published work on CXCR4 antagonists, we have synthesized a series of aryl sulfonamides that inhibit the CXCR4/CXCL12 interaction. Analogue bioactivities were assessed with binding affinity and Matrigel invasion assays. Computer modeling was employed to evaluate a selection of the new analogues docked into the CXCR4 X-ray structure and to rationalize discrepancies between the affinity and Matrigel in vitro assays. A lead compound displays nanomolar potency in the binding affinity assay (IC(50)=8.0 nM) and the Matrigel invasion assay (100 % blockade of invasion at 10 nM). These data demonstrate that benzenesulfonamides are a unique class of CXCR4 inhibitors with high potency.
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Pyridine hydrochloride, 98%
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Benzenesulfonamide, ≥98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
4-(Bromomethyl)benzenesulfonyl chloride, 95%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ≥99.5% (GC)
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%