Merck
  • Respiratory toxicity of buprenorphine results from the blockage of P-glycoprotein-mediated efflux of norbuprenorphine at the blood-brain barrier in mice.
Respiratory toxicity of buprenorphine results from the blockage of P-glycoprotein-mediated efflux of norbuprenorphine at the blood-brain barrier in mice.
Critical care medicine (2012-09-15)
Hisham Alhaddad, Salvatore Cisternino, Xavier Declèves, Nicolas Tournier, Joel Schlatter, Fouad Chiadmi, Patricia Risède, Maria Smirnova, Capucine Besengez, Jean-Michel Scherrmann, Frédéric J Baud, Bruno Mégarbane
ABSTRACT
Deaths due to asphyxia as well as following acute poisoning with severe respiratory depression have been attributed to buprenorphine in opioid abusers. However, in human and animal studies, buprenorphine exhibited ceiling respiratory effects, whereas its metabolite, norbuprenorphine, was assessed as being a potent respiratory depressor in rodents. Recently, norbuprenorphine, in contrast to buprenorphine, was shown in vitro to be a substrate of human P-glycoprotein, a drug-transporter involved in all steps of pharmacokinetics including transport at the blood-brain barrier. Our objectives were to assess P-glycoprotein involvement in norbuprenorphine transport in vivo and study its role in the modulation of buprenorphine-related respiratory effects in mice. University-affiliated research laboratory, INSERM U705, Paris, France. Wild-type and P-glycoprotein knockout female Friend virus B-type mice. Respiratory effects were studied using plethysmography and the P-glycoprotein role at the blood-brain barrier using in situ brain perfusion. Norbuprenorphine(≥ 1 mg/kg) and to a lesser extent buprenorphine (≥ 10 mg/kg) were responsible for dose-dependent respiratory depression combining increased inspiratory (TI) and expiratory times (TE). PSC833, a powerful P-glycoprotein inhibitor, significantly enhanced buprenorphine-related effects on TI (p < .01) and TE (p < .05) and norbuprenorphine-related effects on minute volume (VE, p < .05), TI, and TE (p < .001). In P-glycoprotein-knockout mice, buprenorphine-related effects on VE (p < .01), TE (p < .001), and TI (p < .05) and norbuprenorphine-related effects on VE (p < .05) and TI (p < .001) were significantly enhanced. Plasma norbuprenorphine concentrations were significantly increased in PSC833-treated mice (p < .001), supporting a P-glycoprotein role in norbuprenorphine pharmacokinetics. Brain norbuprenorphine efflux was significantly reduced in PSC833-treated and P-glycoprotein-knockout mice (p < .001), supporting P-glycoprotein-mediated norbuprenorphine transport at the blood-brain barrier. P-glycoprotein plays a key-protective role in buprenorphine-related respiratory effects, by allowing norbuprenorphine efflux at the blood-brain barrier. Our findings suggest a major role for drug-drug interactions that lead to P-glycoprotein inhibition in buprenorphine-associated fatalities and respiratory depression.
Product Description
Supelco
Norbuprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norbuprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®