Merck
  • Mesoionic heterocyclic compounds as candidate messenger RNA cap analogue inhibitors of the influenza virus RNA polymerase cap-binding activity.
Mesoionic heterocyclic compounds as candidate messenger RNA cap analogue inhibitors of the influenza virus RNA polymerase cap-binding activity.
Antiviral chemistry & chemotherapy (2009-06-02)
Ian Mickleburgh, Feng Geng, Laurence Tiley
ABSTRACT
An unusual feature of influenza viral -messenger RNA (mRNA) synthesis is its dependence upon host cell mRNAs as a source of capped RNA primers. A crucial activity of the influenza polymerase is to steal these primers by binding and cleaving the caps from host mRNAs. The recent structural analysis of the cap-binding fragment of the influenza virus PB2 protein has highlighted the importance of the mesoionic properties of the N7-methylguanine (N(7m)G) component of the mRNA cap in this interaction. A series of mesoionic heterocycles with 5,6-fused ring systems analogous to the N(7m)G component of mRNA cap structures were synthesized and examined for the ability to inhibit the cap-binding activity of the influenza virus RNA polymerase complex using a bead-based in vitro cap-binding assay. None of the compounds tested were able to significantly inhibit binding and subsequent endonucleolytic cleavage of a synthetic radiolabelled capped mRNA substrate by recombinant influenza virus polymerase in vitro. Compounds analogous to the mesoionic N(7m)G component of mRNA cap structures comprise a large class of potential inhibitors of the influenza virus polymerase. Although this preliminary assessment of a small group of related analogues was unsuccessful, further screening of this class of compounds is warranted.
Sigma-Aldrich
7-Methylguanine, ≥98.0% (HPLC)