Merck
  • High-performance liquid chromatographic determination of acrolein as a marker for cyclophosphamide bioactivation in human liver microsomes.
High-performance liquid chromatographic determination of acrolein as a marker for cyclophosphamide bioactivation in human liver microsomes.
Journal of chromatography. B, Biomedical sciences and applications (1997-04-25)
F Bohnenstengel, M Eichelbaum, E Golbs, H K Kroemer
ABSTRACT
A high-performance liquid chromatographic method for the quantification of acrolein following incubation of cyclophosphamide (CP) with human liver microsomes was developed. Based on the formation of the fluorescent derivative 7-hydroxyquinoline by condensation of acrolein with 3-aminophenol quantitation was performed without prior extraction or other sample cleanup procedures. The method showed sufficient sensitivity with a limit of detection of 5 ng/ml and a limit of quantification of 10 ng/ml. The suitability of the method is shown for enzyme kinetic studies.
Product Description
Supelco
3-Aminophenol, PESTANAL®, analytical standard
Sigma-Aldrich
3-Aminophenol, 98%