Merck
The binding of platinum complexes to tuna cytochrome c.
The Biochemical journal (1982-03-01)
A P Boswell, G R Moore, R J Williams
ABSTRACT
The binding of [PtCl4]2- and cis-[PtCl2(NH3)2] to methionine-65 of tuna cytochrome c was investigated by 1H n.m.r. The modification at methionine-65 is shown to cause an extremely small structural perturbation to the protein at the site of modification.
Sigma-Aldrich
Platinum(IV) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Platinum(IV) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum(IV) chloride, 96%