Merck
  • Synthesis and reactivity of Zn-biphenyl metal-organic frameworks, introducing a diphenylphosphino functional group.
Synthesis and reactivity of Zn-biphenyl metal-organic frameworks, introducing a diphenylphosphino functional group.
Chimia (2013-08-16)
Marco Ranocchiari, Jeroen Anton van Bokhoven
ABSTRACT
Structural features, synthesis, and reactivity of Zn-biphenyl metal-organic frameworks with MOF-5 topology are presented to show the chemical flexibility of such materials and to demonstrate the challenges that can be encountered and solved to avoid interpenetration. We introduce the synthesis of a Zn-biphenyl MOF with diphenylphosphino functionalization and illustrate its structural and chemical properties.
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ≥99%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, mossy, ≥99%