Merck
  • [Dependence of the operation stress degree from the kind of operative intervention for an acute cholecystitis in the patients with high operative-anesthesiological risk].
[Dependence of the operation stress degree from the kind of operative intervention for an acute cholecystitis in the patients with high operative-anesthesiological risk].
Klinichna khirurhiia (2013-05-31)
M V Bezruchko, S V Malyk, S P Kravchenko, O S Osipov, D A Sytnik
ABSTRACT
The results of comparison between the operation stress degree in various kinds of surgical interventions, performed for an acute cholecystitis, using determination of cortizol, prolactin and glucose content before the operation, intraoperatively and postoperatively in 50 patients, are adduced. There was established, that the largest (in 5.3 times) and the most durable (more than 24 hours) intr erative raising of the cortizol level in the blood serum was noted in patients, to whom open cholecystectomy (OCH) was done, and the minimal (in 2.2 times) and the least durable (up to 1 hour)--while performing transcutaneous transhepatic draining (TTD) of gallbladder under ultrasonographic control. While performance of laparoscopic cholecystectomy (LCH) there was noted the most pronounced intraoperative raising of prolactin level (in 3.6 times) and more rapid its lowering (during 24 hours) in comparison with such while the OCH performance (during 72 hours). In TTD there was observed the minimal intraoperative inhancing of the prolactin level (in 2.3 times) and its duration (during 1 hour) postoperatively. The above mentioned have witnessed, that while TTD of gallbladder performance stimulation of the anterior hypophysis is significantly lesser, than while LCH and OCH.