Merck
  • A preliminary evaluation into the performance of posterior resin bonded cast metal restorations (adhesive onlays).
A preliminary evaluation into the performance of posterior resin bonded cast metal restorations (adhesive onlays).
The European journal of prosthodontics and restorative dentistry (2013-05-21)
S M Marchan, A Eder, Q M Marchan, L Coldero, A Tang Choon, W A Smith
ABSTRACT
Posterior resin bonded cast metal restorations (adhesive onlays) were used in a variety of clinical scenarios including: management of tooth wear and cracked tooth, as retainers for fixed bridge work, for correction of the occlusal plane and in providing cuspal coverage following endodontic treatment. The mean length in service for the examined onlays was 42 months, with a range of 9-75 months. Two restorations in two patients failed resulting in an overall success of 94%. Patient satisfaction was high at 95%. Such restorations seem to be a viable option for managing a number of clinical scenarios.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis