Merck
  • Polyethylene glycol-modified gelatin/polylactic acid nanoparticles for enhanced photodynamic efficacy of a hypocrellin derivative in vitro.
Polyethylene glycol-modified gelatin/polylactic acid nanoparticles for enhanced photodynamic efficacy of a hypocrellin derivative in vitro.
Journal of biomedical nanotechnology (2013-05-01)
Anish Babu, Jayaprakash Periasamy, Amsaveni Gunasekaran, Ganesan Kumaresan, Selvaraj Naicker, Paramasamy Gunasekaran, Ramachandran Murugesan
ABSTRACT
The present study focused on the development of a novel biodegradable nanoparticle system based on polyethyleneglycol-modified gelatin (PEG-GEL) and polylactic acid (PLA) biopolymers for improving the photodynamic efficacy of cyclohexane-1,2-diamino hypocrellin B (CHA2HB), a potent photodynamic therapeutic (PDT) agent. The CHA2HB-loaded PEG-GEL/PLA nanoparticles showed near-spherical morphology with an average size of 190 nm at a PLA to PEG-GEL ratio of 1:3. The drug loading was sufficient enough to produce potentially toxic reactive oxygen species (ROS) needed for photodynamic therapy (PDT). Slow and controlled drug release was observed in normal conditions, whereas enzyme assistance resulted in a relatively fast release due to partial disintegration of CHA2HB-loaded PEG-GEL/PLA nanoparticles. In vitro experiments indicated that CHA2HB-loaded PEG-GEL-PLA nanoparticles are efficiently taken up by cancer cells such as human breast adenocarcinoma (MCF-7), human gastric sarcoma (AGS) and mice specific Dalton's lymphoma (DLA) in a time dependent manner. Further, CHA2HB-loaded PEG-GEL/PLA nanoparticles evoked superior phototoxicity compared to free-CHA2HB towards all the three cell lines investigated. Interestingly, PDT effectiveness was different for the different cell type studied. CHA2HB-loaded PEG-GEL/PLA nanoparticles induced both apoptotic and necrotic cell death as a result of photoirradiation. Thus, our data suggest that PEG-GEL/PLA nanoparticles are highly effective in delivery and phototoxic enhancement of CHA2HB against model cancer cells in vitro.
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture