Merck
  • Identification of two distinct carcinoma-associated fibroblast subtypes with differential tumor-promoting abilities in oral squamous cell carcinoma.
Identification of two distinct carcinoma-associated fibroblast subtypes with differential tumor-promoting abilities in oral squamous cell carcinoma.
Cancer research (2013-04-20)
Daniela Elena Costea, Allison Hills, Amani H Osman, Johanna Thurlow, Gabriela Kalna, Xiaohong Huang, Claudia Pena Murillo, Himalaya Parajuli, Salwa Suliman, Keerthi K Kulasekara, Anne Chr Johannessen, Max Partridge
ABSTRACT
Heterogeneity of carcinoma-associated fibroblasts (CAF) has long been recognized, but the functional significance remains poorly understood. Here, we report the distinction of two CAF subtypes in oral squamous cell carcinoma (OSCC) that have differential tumor-promoting capability, one with a transcriptome and secretome closer to normal fibroblasts (CAF-N) and the other with a more divergent expression pattern (CAF-D). Both subtypes supported higher tumor incidence in nonobese diabetic/severe combined immunodeficient (NOD/SCID) Ilγ2(null) mice and deeper invasion of malignant keratinocytes than normal or dysplasia-associated fibroblasts, but CAF-N was more efficient than CAF-D in enhancing tumor incidence. CAF-N included more intrinsically motile fibroblasts maintained by high autocrine production of hyaluronan. Inhibiting CAF-N migration by blocking hyaluronan synthesis or chain elongation impaired invasion of adjacent OSCC cells, pinpointing fibroblast motility as an essential mechanism in this process. In contrast, CAF-D harbored fewer motile fibroblasts but synthesized higher TGF-β1 levels. TGF-β1 did not stimulate CAF-D migration but enhanced invasion and expression of epithelial-mesenchymal transition (EMT) markers in malignant keratinocytes. Inhibiting TGF-β1 in three-dimensional cultures containing CAF-D impaired keratinocyte invasion, suggesting TGF-β1-induced EMT mediates CAF-D-induced carcinoma cell invasion. TGF-β1-pretreated normal fibroblasts also induced invasive properties in transformed oral keratinocytes, indicating that TGF-β1-synthesizing fibroblasts, as well as hyaluronan-synthesizing fibroblasts, are critical for carcinoma invasion. Taken together, these results discern two subtypes of CAF that promote OSCC cell invasion via different mechanisms.
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide