Merck
  • The therapeutic efficacy of propranolol in children with recurrent primary epistaxis.
The therapeutic efficacy of propranolol in children with recurrent primary epistaxis.
Drug design, development and therapy (2013-03-08)
Bojko Bjelakovic, Mila Bojanovic, Stevo Lukic, Ljiljana Saranac, Vladislav Vukomanovic, Sergej Prijic, Nikola Zivkovic, Dusica Randjelovic
ABSTRACT
We hypothesized that some characteristics of beta-blockers, including negative inotropic, peripheral vasoconstrictor, and antiangiogenic effects, might be potentially useful in treating children with epistaxis. From June 2010 to March 2012, a total of seven children with recurrent primary epistaxis resistant to conventional management were observed at our institution. An overall effectiveness of propranolol was noted in all seven children when given a dose of 1.5-2 mg/kg/day (divided into three doses) as a second line therapy for terminating epistaxis. Based on our first experience, we believe that propranolol could be a favorable treatment option for patients with primary epistaxis.
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Supelco
Propranolol hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard