Merck
  • Anti-dyskinetic effect of the neuronal nitric oxide synthase inhibitor is linked to decrease of FosB/deltaFosB expression.
Anti-dyskinetic effect of the neuronal nitric oxide synthase inhibitor is linked to decrease of FosB/deltaFosB expression.
Neuroscience letters (2013-02-23)
Fernando Eduardo Padovan-Neto, Nádia Rubia Ferreira, Danielle de Oliveira-Tavares, Daniele de Aguiar, Célia Aparecida da Silva, Rita Raisman-Vozari, Elaine Del Bel
ABSTRACT
Rodents with lesion of dopaminergic pathway when receiving repeated l-3,4-dihydroxiphenylalanine (l-DOPA) treatment develop abnormal involuntary movements called dyskinesia. We demonstrated that nitric oxide synthase (NOS) inhibitors mitigate l-DOPA-induced dyskinesia in rodents. The aim of the present study was to verify if the in vivo preferential neuronal NOS (nNOS) inhibitor 7-nitroindazole (7-NI) affect the expression of the transcription factor FosB/ΔFosB in the lesioned striatum, an indicator of neuronal activity associated with dyskinesia. Male Wistar rats with unilateral microinjection (medial forebrain bundle) of either the neurotoxin 6-hydroxidopamine (6-OHDA; n=4-6/group) or saline (sham; n=6/group) were provided with l-DOPA (30mg/kg plus benserazide 7.5mg/kg/day, oral gavage), once a day during 22 days. 6-OHDA-lesioned animals developed abnormal involuntary movements (AIMs) classified as axial, limb, orofacial and locomotive dyskinesia and presented FosB/ΔFosB increase in the dopamine-depleted striatum. Administration of 7-NI (30mg/kg, i.p.), 30min prior to l-DOPA reduced the severity of AIMs (≈65% for axial, limb and orofacial and 74% for locomotive AIMs scores), without interfering with the rotarod performance. Simultaneously, 7-NI attenuated the expression of FosB/ΔFosB in dopamine-depleted striatum (≈65% in medial and ≈54% in lateral striatum, bregma 0.48mm). FosB/ΔFosB expression in lateral striatum was correlated with l-DOPA-induced dyskinesia. The findings described here corroborate a new approach to the management of l-DOPA-therapy in Parkinson's disease (PD) treatment.
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder
Sigma-Aldrich
6-Hydroxydopamine hydrobromide, 95%
Sigma-Aldrich
7-Nitroindazole, ≥98%