Merck
  • Pyramidal neurons derived from human pluripotent stem cells integrate efficiently into mouse brain circuits in vivo.
Pyramidal neurons derived from human pluripotent stem cells integrate efficiently into mouse brain circuits in vivo.
Neuron (2013-02-12)
Ira Espuny-Camacho, Kimmo A Michelsen, David Gall, Daniele Linaro, Anja Hasche, Jérôme Bonnefont, Camilia Bali, David Orduz, Angéline Bilheu, Adèle Herpoel, Nelle Lambert, Nicolas Gaspard, Sophie Péron, Serge N Schiffmann, Michele Giugliano, Afsaneh Gaillard, Pierre Vanderhaeghen
ABSTRACT
The study of human cortical development has major implications for brain evolution and diseases but has remained elusive due to paucity of experimental models. Here we found that human embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs), cultured without added morphogens, recapitulate corticogenesis leading to the sequential generation of functional pyramidal neurons of all six layer identities. After transplantation into mouse neonatal brain, human ESC-derived cortical neurons integrated robustly and established specific axonal projections and dendritic patterns corresponding to native cortical neurons. The differentiation and connectivity of the transplanted human cortical neurons complexified progressively over several months in vivo, culminating in the establishment of functional synapses with the host circuitry. Our data demonstrate that human cortical neurons generated in vitro from ESC/iPSC can develop complex hodological properties characteristic of the cerebral cortex in vivo, thereby offering unprecedented opportunities for the modeling of human cortex diseases and brain repair.
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.