Merck
Colloidal aggregation in microgravity by critical Casimir forces.
Physical review letters (2013-02-02)
Sandra J Veen, Oleg Antoniuk, Bart Weber, Marco A C Potenza, Stefano Mazzoni, Peter Schall, Gerard H Wegdam
ABSTRACT
By using the critical Casimir force, we study the attractive strength dependent aggregation of colloids with and without gravity by means of near field scattering. Significant differences were seen between microgravity and ground experiments, both in the structure of the formed fractal aggregates as well as in the kinetics of growth. In microgravity purely diffusive aggregation is observed. By using the continuously variable particle interaction potential we can for the first time experimentally relate the strength of attraction between the particles and the structure of the aggregates.
Sigma-Aldrich
3-Methylpyridine, 99%
Sigma-Aldrich
3-Methylpyridine, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D