Merck
  • New generation of efficient peptide-based vectors, NickFects, for the delivery of nucleic acids.
New generation of efficient peptide-based vectors, NickFects, for the delivery of nucleic acids.
Biochimica et biophysica acta (2013-01-30)
Piret Arukuusk, Ly Pärnaste, Nikita Oskolkov, Dana-Maria Copolovici, Helerin Margus, Kärt Padari, Kaidi Möll, Julia Maslovskaja, Radi Tegova, Gaily Kivi, Andres Tover, Margus Pooga, Mart Ustav, Ulo Langel
ABSTRACT
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting oligonucleotides and short interfering RNAs into stable nanoparticles with a size of 62-160nm. Such nanoparticles have a negative surface charge (-11 to -18mV) in serum containing medium and enable highly efficient gene expression, splice correction and gene silencing. One of the novel peptides, NickFect51 is capable of transfecting plasmid DNA into a large variety of cell lines, including refractory suspension and primary cells and in several cases exceeds the transfection level of commercially available reagent Lipofectamine™ 2000 without any cytotoxic side effects. Additionally we demonstrate the advantages of NickFect51 in a protein production system, QMCF technology, for expression and production of recombinant proteins in hardly transfectable suspension cells.
Product Description
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)