Merck
  • Influence of nitrogen excited species on the destruction of naphthalene in nitrogen and air using surface dielectric barrier discharge.
Influence of nitrogen excited species on the destruction of naphthalene in nitrogen and air using surface dielectric barrier discharge.
Journal of hazardous materials (2013-01-03)
Ayman A Abdelaziz, Takafumi Seto, M Abdel-Salam, Yoshio Otani
ABSTRACT
The destruction of naphthalene, as representative polycyclic aromatic hydrocarbons, by surface dielectric barrier discharge is investigated in air as well as dry and humidified nitrogen at ambient temperature. Naphthalene destruction efficiency is evaluated in terms of chemical change vis-a-vis energy utilization. The detected byproducts are qualitatively evaluated in order to understand the role of the active species in the destruction process. The results show that the destruction efficiency and the energy efficiency are higher in the dry nitrogen than in the humidified nitrogen, and these decrease with the increase of the humidity. Measured concentration of ozone as a byproduct qualitatively indicates the roles of oxygen and ozone in the destruction process in air. The analysis of the aerosol particles formed during the destruction process, both in the dry and humidified nitrogen, confirmed the adverse effects of the humidity on the byproducts formation and subsequently on the destruction process.
Product Description
Supelco
Melting point standard 79-81°C, analytical standard
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland