Merck
  • Synthesis and preliminary evaluation of pharmacological properties of some piperazine derivatives of xanthone.
Synthesis and preliminary evaluation of pharmacological properties of some piperazine derivatives of xanthone.
Bioorganic & medicinal chemistry (2012-12-19)
Natalia Szkaradek, Anna Rapacz, Karolina Pytka, Barbara Filipek, Agata Siwek, Marek Cegła, Henryk Marona
ABSTRACT
A series of 9 piperazine derivatives of xanthone were synthesized and evaluated for cardiovascular activity. The following pharmacological experiments were conducted: the binding affinity for adrenoceptors, the influence on the normal electrocardiogram, the effect on the arterial blood pressure and prophylactic antiarrhythmic activity in adrenaline induced model of arrhythmia (rats, iv). Three compounds revealed nanomolar affinity for α(1)-adrenoceptor which was correlated with the strongest cardiovascular (antiarrhythmic and hypotensive) activity in animals' models. The most promising compound was 4-(3-(4-(2-methoxyphenyl)piperazine-1-yl)propoxy)-9H-xanthen-9-one hydrochloride (12) which revealed antiarrhythmic activity with ED(50) value of 0.69 mg/kg in adrenaline induced arrhythmia (rats, iv). Other synthesized xanthone derivatives, that is, (R,S)-4-(2-hydroxy-3-(4-(2-methoxyphenyl)piperazine-1-yl)propoxy)-9H-xanthen-9-one hydrochloride (10) and (R,S)-4-(2-acetoxy-3-(4-(2-methoxyphenyl)piperazine-1-yl)propoxy)-9H-xanthen-9-one hydrochloride (11) also acted as potential antiarrhythmics in adrenaline induced model of arrhythmia in rats after intravenous injection (ED(50) = 0.88 mg/kg and 0.89 mg/kg, respectively). These values were lower than values obtained for reference drugs such as propranolol and urapidil, but not carvedilol. Results were quite promising and suggested that in the group of xanthone derivatives new potential antiarrhythmics and hypotensives might be found.
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Sigma-Aldrich
Xanthone, 97%
Supelco
Piperazine hexahydrate, analytical standard