Merck
  • Differential metabolite profiles and salinity tolerance between two genetically related brown-seeded and yellow-seeded Brassica carinata lines.
Differential metabolite profiles and salinity tolerance between two genetically related brown-seeded and yellow-seeded Brassica carinata lines.
Plant science : an international journal of experimental plant biology (2012-12-04)
Thomas Canam, Xiang Li, Jennifer Holowachuk, Min Yu, Jianguo Xia, Rupasri Mandal, Ramanarayan Krishnamurthy, Souhaila Bouatra, Igor Sinelnikov, Bianyun Yu, Larry Grenkow, David S Wishart, Harold Steppuhn, Kevin C Falk, Tim J Dumonceaux, Margaret Y Gruber
ABSTRACT
Brassica carinata (Ethiopian mustard) has previously been identified as a potential crop species suitable for marginal land in the North American prairies due to its relatively high salt tolerance. Two genetically related B. carinata lines with brown-seeded (BS) and yellow-seeded (YS) phenotypes were assessed for their tolerance to sodium sulfate. Specifically, each line was greenhouse-grown under 0, 50 and 100mM of salt, and analyzed after four weeks and eight weeks of treatment. Generally, the height of the BS line was greater than the YS line under both salt treatments, indicating enhanced salt tolerance of the BS line. NMR-based metabolite profiling and PCA analyses indicated a more pronounced shift in key stem metabolites after four weeks of treatment with the YS line compared to the BS line. For example, tryptophan and formate levels increased in the YS line after four weeks of 100mM salt treatment, while proline and threonine levels varied uniquely compared to other metabolites of the lines. Together, the data indicate that the brown-seeded line has greater sodium tolerance than the yellow-seed line, provide clues to the biochemical underpinnings for the phenotypic variation, and highlight the utility of B. carinata as a biorefinery crop for saline land.
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%