Merck
  • A new three-dimensional metal-organic framework constructed from 9,10-anthracene dibenzoate and Cd(II) as a highly active heterogeneous catalyst for oxidation of alkylbenzenes.
A new three-dimensional metal-organic framework constructed from 9,10-anthracene dibenzoate and Cd(II) as a highly active heterogeneous catalyst for oxidation of alkylbenzenes.
Dalton transactions (Cambridge, England : 2003) (2012-10-20)
Dabin Shi, Yanwei Ren, Huanfeng Jiang, Jiaxian Lu, Xiaofei Cheng
ABSTRACT
A novel three-dimensional (3D) porous metal-organic framework, {[Cd(L)(H(2)O)]·3H(2)O}(∞) (1) (L-H(2) = 4,4'-(9,10-anthracenediyl)dibenzoic acid), was synthesized. 1 has a 3D framework formed by L connectors and the infinite {Cd(O(2)CR)(2)}(∞) secondary building units (SBUs). Compound 1 was characterized by IR spectroscopy, thermogravimetry, single-crystal, elemental analyses and powder X-ray diffraction methods. We then investigated its adsorption, catalysis and luminescent properties. Framework 1' exhibits permanent porosity (Langmuir surface area, 324 m(2) g(-1)), high thermal stability (up to 380 °C), and highly active properties for oxidation of alkylbenzenes. The catalytic results reveal that 1' is indeed an efficient heterogeneous catalyst with 100% selectivity. To the best of our knowledge, it is the first report that oxidation reactions of alkylbenzenes were performed using Cd(II) with unsaturated coordination spheres as active sites.
Product Description
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)