Merck
  • Decolourization of the azo dye Orange G in aqueous solution via a heterogeneous Fenton-like reaction catalysed by goethite.
Decolourization of the azo dye Orange G in aqueous solution via a heterogeneous Fenton-like reaction catalysed by goethite.
Environmental technology (2012-09-20)
Honghai Wu, Xiaowen Dou, Dayi Deng, Yufeng Guan, Liguo Zhang, Guangping He
ABSTRACT
Decolourization of the azo dye Orange G (OG) was investigated by using goethite/H2O2 as a heterogeneous Fenton-like reagent. Five principle operational parameters, namely pH, ion strength, concentrations ofgoethite (alpha-FeOOH) and hydrogen peroxide (H2O2), and reaction temperature, were taken into account to investigate how these controlling factors mediated OG decolourization. Goethite surfaces catalysed a Fenton-like reaction responsible for decolourizing OG following pseudo-first-order kinetics (R2 > 0.964). This process was effective but seriously impacted by the medium pH and the dosages of both alpha-FeOOH and H2O2. The decolourization efficiencies of OG increased with the decrease of solution pH and NaCl (chloride ion) concentration and/or the increase of H2O2. The acidic aqueous medium conditions were likely favourable due to the surface adsorption of the negatively charged OG leading to the promotion of decolourizing OG. The apparent activation energy (E) for this reaction was 42.18 kJ mol(-1), a relatively low value. This is consistent with the OG decolourization being enhanced with the reaction temperature increase.
Sigma-Aldrich
Orange G, certified by the Biological Stain Commission, Dye content 80 %
Sigma-Aldrich
Orange G, for NA electrophoresis
Sigma-Aldrich
Orange G, certified by the Biological Stain Commission