Merck
  • Polymer coating of carrier excipients modify aerosol performance of adhered drugs used in dry powder inhalation therapy.
Polymer coating of carrier excipients modify aerosol performance of adhered drugs used in dry powder inhalation therapy.
International journal of pharmaceutics (2012-09-12)
Daniela Traini, Santo Scalia, Handoko Adi, Elisabetta Marangoni, Paul M Young
ABSTRACT
The potential of excipient coating to enhance aerosol performance of micronized drugs in carrier excipient-drug blends, used in dry powder inhalers, was investigated. Both EC (ethyl cellulose) and PVP (polyvinylpyrrolidone) were used as coating agents. Carriers were prepared via sieve fractioning followed by spray drying, with and without polymer additive. Each uncoated and coated carrier salbutamol sulphate (SS) blended systems were evaluated for particle size, morphology, drug carrier adhesion and aerosolisation performance, after blending and storage for 24h. All carrier-based systems prepared had similar particle sizes and morphologies. The surface chemistries of the carriers were significantly different, as was drug-carrier adhesion and aerosolisation performance. Particle adhesion between SS and aerosol performance (fine particle fraction; FPF) followed the rank: PVP coated>un-coated>EC coated lactose. This rank order could be attributed to the surface energy measured by contact goniometry and related to the chemistry of lactose and each polymer. Storage did not significantly affect aerosol performance, however a rank increase in mean FPF value was observed for uncoated and EC coated lactose. Finally, the net electrostatic charge across the aerosol cloud indicated that the EC coated lactose transferred less charge to SS particles. The performance of each carrier system could be attributed to the carrier surface chemistry and, in general, by careful selection of the coating polymer, drug-carrier adhesion, electrostatic charge and aerosol performance could be controlled.
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis