Merck
  • Improved chemical stabilities for end-capped high performance liquid chromatography stationary phases based on poly(methyloctadecylsiloxane) thermally immobilized onto metalized silicas.
Improved chemical stabilities for end-capped high performance liquid chromatography stationary phases based on poly(methyloctadecylsiloxane) thermally immobilized onto metalized silicas.
Journal of chromatography. A (2012-09-12)
Carla G A da Silva, Carol H Collins
ABSTRACT
Endcapped stationary phases were prepared after thermal immobilization of poly(methyloctadecylsiloxane) (PMODS) onto zirconized and titanized silica supports. These new stationary phases have lower densities of residual hydroxyl groups, according to infrared spectroscopy and 29Si CP-MAS NMR and as shown by the symmetrical peaks of basic compounds from the Tanaka, Engelhardt and SRM 870 test mixtures. Stability tests for the endcapped stationary phases, measured using severe alkaline conditions (70:30 (v/v) methanol:0.05 mol/L K2CO3/KHCO3, pH 10, 50 °C), revealed that the stabilities of these phases are greater than the stabilities of similar nonendcapped phases. The stationary phases showed good performance for the separation of basic pharmaceuticals.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D)
Supelco
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%