Merck
  • Engineering surface adhered poly(vinyl alcohol) physical hydrogels as enzymatic microreactors.
Engineering surface adhered poly(vinyl alcohol) physical hydrogels as enzymatic microreactors.
ACS applied materials & interfaces (2012-09-04)
Betina Fejerskov, Bettina E B Jensen, Najah B S Jensen, Siow-Feng Chong, Alexander N Zelikin
ABSTRACT
In this work, we characterize physical hydrogels based on poly(vinyl alcohol), PVA, as intelligent biointerfaces for surface-mediated drug delivery. Specifically, we assemble microstructured (μS) surface adhered hydrogels via noncryogenic gelation of PVA, namely polymer coagulation using sodium sulfate (Na(2)SO(4)). We present systematic investigation of concentrations of Na(2)SO(4) as a tool of control over assembly of μS PVA hydrogels and quantify polymer losses and retention within the hydrogels. For polymer quantification, we use custom-made PVA with single terminal thiol group in a form of mixed disulfide with Ellman's reagent which provides for a facile UV-vis assay of polymer content in coagulation baths, subsequent washes in physiological buffer, and within the hydrogel phase. Polymer coagulation using varied concentrations of sodium sulfate afforded biointerfaces with controlled elasticity for potential uses in investigating mechano-sensitive effects of mammalian cell culture. For surface mediated drug delivery, we propose a novel concept termed Substrate Mediated Enzyme Prodrug Therapy (SMEPT) and characterize μS PVA hydrogels as reservoirs for enzymatic cargo. Assembled functional interfaces are used as matrices for cell culture and delivery of anticancer drug achieved through administration of a benign prodrug, its conversion into an active therapeutic within the hydrogel phase, and subsequent internalization by adhered hepatic cells. Taken together, the presented data contribute significantly to the development of novel matrices for surface-mediated drug delivery and other biomedical applications.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%