Merck
  • Drag-reducing polymers: a novel class of diuretic and natriuretic compounds.
Drag-reducing polymers: a novel class of diuretic and natriuretic compounds.
Cardiovascular drugs and therapy (1990-02-01)
D D Smyth, P I Polimeni
ABSTRACT
Previous studies have demonstrated that Separan AP-30, a drag-reducing polymer, significantly decreased the formation of atherosclerotic plaques in rabbits fed a high-cholesterol diet. Furthermore, Separan AP-273, a polymer similar to but longer than Separan AP-30, markedly increased cardiac output in open-chest rats. Preliminary studies in our laboratory have described diuresis and natriuresis following the intravenous infusion of Separan AP-273. These studies did not determine whether this natriuresis was common to drag-reducing polymers or unique to Separan. In the present study the renal effects of two chemically dissimilar drag-reducing polymers of molecular lengths approaching approximately 100 microns were compared. Sprague-Dawley rats (250-300 g) were anesthetized (Nembutal) and a tracheostomy was performed. A baseline level of sodium and water excretion was established by the infusion of saline (0.097 ml/min, IV). Separan (0.004%), an anionic polyacrylamide; Polyox (0.4%), a poly(ethylene oxide); or mannitol (20%), an osmotic diuretic, was infused into the jugular vein at 0.0034 ml/min. Separan and Polyox both increased the excretion of sodium and water (p less than 0.05). At the doses tested, potassium excretion was unaltered in both groups. Separan, but not Polyox, produced a modest increase in blood pressure. Creatinine clearance was unaltered. At this infusion rate, only hypertonic concentrations of mannitol (20%) induced a similar effect. These results indicate that drag-reducing polymers may represent a novel class of potassium-sparing diuretic and natriuretic compounds. The ability of this class of polymers to decrease the formation of atherosclerotic plaques, increase blood flow, and produce a potassium-sparing natriuresis may be of practical clinical significance.
Product Description
Supelco
Polyethylene glycol/Polyethylene oxide ReadyCal Set Mp ~250 -1′100 000, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Kollisolv® PEG E 300
Pricing and availability is not currently available.
Sigma-Aldrich
Kollisolv® PEG E 400
Pricing and availability is not currently available.
Sigma-Aldrich
Polyethylene glycol/ dimethyl sulfoxide solution, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), average Mn 4,600
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~8,000,000 (nominal), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~2,000,000 (nominal), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), average Mn 950-1,050
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 1,450
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), Hybri-Max, mol wt 1,300-1,600, waxy solid, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), Hybri-Max, mol wt 3,000-3,700, waxy solid, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,350
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), average Mn 10,000, flakes
Pricing and availability is not currently available.
Sigma-Aldrich
Polyethylene glycol 3000 monodisperse solution, BioUltra, for molecular biology, ~50% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, for molecular biology, 1,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, for molecular biology, 6,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 6,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 2,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 8,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 1,500
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 3,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 40,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, for molecular biology, 8,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 4,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 400
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 600
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 108,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 12,000
Pricing and availability is not currently available.
Supelco
Poly(ethylene glycol), analytical standard, for GPC, 20,000
Pricing and availability is not currently available.