Merck
  • Low molecular mass chitosan as carrier for a hydrodynamically balanced system for sustained delivery of ciprofloxacin hydrochloride.
Low molecular mass chitosan as carrier for a hydrodynamically balanced system for sustained delivery of ciprofloxacin hydrochloride.
Acta pharmaceutica (Zagreb, Croatia) (2012-07-04)
Anurag Verma, Ashok K Bansal, Amitava Ghosh, Jayanta K Pandit
ABSTRACT
Chitosan has become a focus of major interest in recent years due to its excellent biocompatibility, biodegradability and non-toxicity. Although this material has already been extensively investigated in the design of different types of drug delivery systems, it is still little explored for stomach specific drug delivery systems. The objective of the present investigation was to explore the potential of low molecular mass chitosan (LMCH) as carrier for a hydrodynamically balanced system (HBS) for sustained delivery of water soluble drug ciprofloxacin hydrochloride (CP). Various formulations were prepared by physical blending of drug and polymer(s) in varying ratios followed by encapsulation into hard gelatin capsules. All the formulations remained buoyant in 0.1 mol L⁻¹ HCl (pH 1.2) throughout the experiment. Effect of addition of xanthan gum (XG) or ethyl cellulose (EC) on drug release was also investigated. Zero order drug release was obtained from the formulations containing LMCH alone or in combination with XG, and in one instance also with EC. Our results suggest that LMCH alone or in combination with XG is an excellent material for stomach specific sustained delivery of CP from hydrodynamically balanced single unit capsules.
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Ethyl cellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethyl cellulose, 48.0-49.5% (w/w) ethoxyl basis