Merck
  • Interactions of D-cellobiose with p-toluenesulfonic acid in aqueous solution: a (13)C NMR study.
Interactions of D-cellobiose with p-toluenesulfonic acid in aqueous solution: a (13)C NMR study.
Carbohydrate research (2011-11-01)
Ananda S Amarasekara, Onome S Owereh, Brian Ezeh
ABSTRACT
The effects of adding D(2)SO(4), and p-toluenesulfonic acid-d to D-cellobiose dissolved in D(2)O were investigated at 23°C by plotting (13)C NMR chemical shift changes (Δδ) against the acid to D-cellobiose molar ratio. (13)C Chemical shifts of all 18 carbon signals from α and β anomers of D-cellobiose showed gradual decreases due to increasing acidity in aqueous D(2)SO(4) medium. The C-1 of the α anomer showed a slightly higher response to increasing D(+) concentration in the surrounding. In the aqueous p-toluenesulfonic acid-d medium, C-6' and C-4' carbons of both α, and β anomeric forms of D-cellobiose are significantly affected by increasing the sulfonic acid concentrations, and this may be due to a 1:1 interaction of p-toluenesulfonic acid-d with the C-6', C-4' region of the cellobiose molecule.
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium p-toluenesulfonate, 95%
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ReagentPlus®, ≥98%
Sigma-Aldrich
Silver p-toluenesulfonate, ≥99%