Merck
  • Morphological changes of the filamentous fungus Mucor mucedo and inhibition of chitin synthase activity induced by anethole.
Morphological changes of the filamentous fungus Mucor mucedo and inhibition of chitin synthase activity induced by anethole.
Phytotherapy research : PTR (2011-07-02)
Masahiro Yutani, Yukie Hashimoto, Akira Ogita, Isao Kubo, Toshio Tanaka, Ken-ichi Fujita
ABSTRACT
trans-Anethole (anethole), a major component of anise oil, has a broad antimicrobial spectrum with antimicrobial activity relatively weaker than those of well-known antibiotics, and significantly enhances the antifungal activity of polygodial and dodecanol against the baker's yeast Saccharomyces cerevisiae and human pathogenic yeast Candida albicans. However, the antifungal mechanism of anethole is unresolved. Anethole demonstrated antifungal activity against the filamentous fungus, Mucor mucedo IFO 7684, accompanied by hyphal morphological changes such as swollen hyphae at the tips. Its minimum growth inhibitory concentration was 0.625 mM. A hyperosmotic condition (1.2 M sorbitol) restricted the induction of morphological changes, while hypoosmotic treatment (distilled water) induced bursting of hyphal tips and leakage of cytoplasmic constituents. Furthermore, anethole dose-dependently inhibited chitin synthase (CHS) activity in permeabilized hyphae in an uncompetitive manner. These results suggest that the morphological changes of M. mucedo could be explained by the fragility of cell walls caused by CHS inhibition.
Product Description
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Supelco
trans-Anethole, analytical standard