Merck
  • Low threshold power density for the generation of frequency up-converted pulses in bismuth glass by two crossing chirped femtosecond pulses.
Low threshold power density for the generation of frequency up-converted pulses in bismuth glass by two crossing chirped femtosecond pulses.
Optics express (2011-07-01)
Hang Zhang, Hui Liu, Jinhai Si, Wenhui Yi, Feng Chen, Xun Hou
ABSTRACT
We investigated the generation of frequency up-converted femtosecond laser pulses by nondegenerate cascaded four-wave mixing (CFWM) in a bismuth-oxide glass (BI glass). Broad-bandwidth light pulses with different propagation directions were simultaneously obtained by using two small-angle crossing femtosecond laser pulses in BI glass. Experimental results show that the threshold power density for the generation of broad-bandwidth femtosecond pulses in BI glass is one order of magnitude lower than that in fused silica.
Sigma-Aldrich
Bismuth(III) oxide, nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, purum, ≥98.0% (KT)
Sigma-Aldrich
Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, powder, 99.999% trace metals basis