Merck
  • Levosulpiride, (S)-(-)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl) methyl]-2-methoxybenzamide, enhances the transduction efficiency of PEP-1-ribosomal protein S3 in vitro and in vivo.
Levosulpiride, (S)-(-)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl) methyl]-2-methoxybenzamide, enhances the transduction efficiency of PEP-1-ribosomal protein S3 in vitro and in vivo.
BMB reports (2011-05-28)
Eun Hee Ahn, Dae Won Kim, Duk-Soo Kim, Su Jung Woo, Hye Ri Kim, Joon Kim, Soon Sung Lim, Tae-Cheon Kang, Dong Joon Kim, Ki Tae Suk, Jinseu Park, Qiuxiang Luo, Won Sik Eum, Hyun Sook Hwang, Soo Young Choi
ABSTRACT
Many proteins with poor transduction efficiency were reported to be delivered to cells by fusion with protein transduction domains (PTDs). In this study, we investigated the effect of levosulpiride on the transduction of PEP-1 ribosomal protein S3 (PEP-1-rpS3), and examined its influence on the stimulation of the therapeutic properties of PEP-1-rpS3. PEP-1-rpS3 transduction into HaCaT human keratinocytes and mouse skin was stimulated by levosulpiride in a manner that did not directly affect the cell viability. Following 12-O-tetradecanoylphorbol- 13-acetate (TPA)-induced inflammation in mice, levosulpiride alone was ineffective in reducing TPA-induced edema and in inhibiting the elevated productions of inflammatory mediators and cytokines, such as cyclooxygenase-2, inducible nitric oxide synthase, interleukin-6 and -1β, and tumor necrosis factor- α. Anti-inflammatory activity by PEP-1-rpS3 + levosulpiride was significantly more potent than by PEP-1-rpS3 alone. These results suggest that levosulpiride may be useful for enhancing the therapeutic effect of PEP-1-rpS3 against various inflammatory diseases. [BMB reports 2011; 44(5): 329-334].
Sigma-Aldrich
(S)-(−)-Sulpiride, ≥98% (titration)
Supelco
(S)-(−)-Sulpiride, VETRANAL®, analytical standard