Merck
  • Precision synthesis of poly(3-hexylthiophene) from catalyst-transfer Suzuki-Miyaura coupling polymerization.
Precision synthesis of poly(3-hexylthiophene) from catalyst-transfer Suzuki-Miyaura coupling polymerization.
Macromolecular rapid communications (2011-04-22)
Tsutomu Yokozawa, Ryosuke Suzuki, Masataka Nojima, Yoshihiro Ohta, Akihiro Yokoyama
ABSTRACT
(t)Bu(3) PPd(Ph)Br (1)-catalyzed Suzuki-Miyaura coupling polymerization of 2-(4-hexyl-5-iodo-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (2) was investigated. Monomer 2 was polymerized with 1 at 0 °C in the presence of CsF and 18-crown-6 in THF containing a small amount of water to yield P3HT with a narrow molecular weight distribution and almost perfect head-to-tail regioregularity. The M(n) values increased up to 11,400 g · mol(-1) in proportion to the feed ratio of 2 to 1. The MALDI-TOF mass spectra showed that P3HT with moderate molecular weight uniformly had a phenyl group at one end and a hydrogen atom at the other, indicating involvement of a catalyst-transfer mechanism. Successive 1-catalyzed polymerization of fluorene monomer 3 and then 2 yielded a well-defined block copolymer of polyfluorene and P3HT.
Sigma-Aldrich
18-Crown-6, ≥99.0%
Sigma-Aldrich
18-Crown-6, 99%
Sigma-Aldrich
18-Crown-6, purum, ≥99.0% (GC)
Sigma-Aldrich
18-Crown-6 solution, 1.0 M in THF