Merck
  • anti-Selective catalytic asymmetric nitroaldol reaction via a heterobimetallic heterogeneous catalyst.
anti-Selective catalytic asymmetric nitroaldol reaction via a heterobimetallic heterogeneous catalyst.
Journal of the American Chemical Society (2009-09-10)
Tatsuya Nitabaru, Akihiro Nojiri, Makoto Kobayashi, Naoya Kumagai, Masakatsu Shibasaki
ABSTRACT
Full details of an anti-selective catalytic asymmetric nitroaldol reaction promoted by a heterobimetallic catalyst comprised of Nd(5)O(O(i)Pr)(13), an amide-based ligand, and NaHMDS (sodium hexamethyldisilazide) are described. A systematic synthesis and evaluation of amide-based ligands led to the identification of optimum ligand 1m, which provided a suitable platform for the Nd/Na heterobimetallic complex. During the catalyst preparation in THF, a heterogeneous mixture developed and centrifugation of the suspension allowed for separation of the precipitate, which contained the active catalyst and which could be stored for at least 1 month without any loss of catalytic performance. The precipitate promoted a nitroaldol (Henry) reaction for a broad range of nitroalkanes and aldehydes under heterogeneous conditions, affording the corresponding 1,2-nitroalkanol in a highly anti-selective (up to anti/syn = >40/1) and enantioselective manner (up to 98% ee). Inductively coupled plasma (ICP) and X-ray fluorescence (XRF) analyses revealed that the precipitate indeed included both neodymium and sodium, which was further supported by high-resolution ESI TOF MS spectrometry.
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide ChemBeads