Merck
  • Design and in vitro evaluation of a novel vaginal drug delivery system based on gelucire.
Design and in vitro evaluation of a novel vaginal drug delivery system based on gelucire.
Current drug delivery (2009-05-20)
M Patel Geeta, M Patel Madhabhai
ABSTRACT
Carbamazepine indicated for the control of epilepsy, undergoes extensive hepatic first-pass metabolism after oral administration. A vaginal dosage form of carbamazepine is not commercially available. Conventional suppository having poor retention in the vaginal tract, as they are removed in a short time by the tract's self-cleansing action, having poor patient compliance. To overcome such problems, delivery system with mucoadhesive polymers polyox WSR N-60K and Ucarflock 302 that prolong drug permanence on the vaginal mucosa were developed. In the present study the suitability of gelucires to formulate vaginal pesseries was investigated. The possible modification of carbamazepine release kinetics by using gelucires blends and hydrophilic additives in the pesseries was evaluated. It was observed that among gelucire grades melting point higher than 37 degrees C, the release rate proved to be highly dependant on HLB value and matrix composition. In most of the formulations carbamazepine release occurred by disintegration and erosion of the matrices which is depending upon the vehicle employed. The aging study revealed that the formulations containing G50/13 and G50/13-G44/14 blends undergo some changes during one year of shelf aging. From the results obtained it can be concluded that different gelucire grades and their blends along with hydrophilic polymer could be successesively used to formulate prolong release carbamazepine pesseries.
Sigma-Aldrich
Kollisolv® PEG E 400
Supelco
Polyethylene glycol/Polyethylene oxide ReadyCal Set Mp ~250 -1′100 000, analytical standard
Sigma-Aldrich
Polyethylene glycol/ dimethyl sulfoxide solution, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Kollisolv® PEG E 300
Sigma-Aldrich
Poly(ethylene glycol), average Mn 600, waxy solid (moist)
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 400
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 35,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 20,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 1,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 1,500
Sigma-Aldrich
Poly(ethylene glycol), 35,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 8,000
Sigma-Aldrich
Polyethylene glycol solution, 40 % (w/w) in H2O, average mol wt 8,000
Sigma-Aldrich
Polyethylene glycol solution, Hybri-Max, 50 % (w/v), average mol wt 1,450, 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Poly(ethylene glycol), Hybri-Max, mol wt 1,300-1,600, waxy solid, BioReagent, suitable for hybridoma
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 10,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 400
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 4,000
Sigma-Aldrich
Poly(ethylene glycol), average Mn 4,000, platelets
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 6,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,350
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 4,000
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 200
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~8,000,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv ~900,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene oxide), average Mv 600,000 (nominal), powder
Sigma-Aldrich
Poly(ethylene glycol), average Mn 4,600
Sigma-Aldrich
Poly(ethylene glycol), average Mn 300
Sigma-Aldrich
Poly(ethylene glycol), average Mv ~8,000, powder (crystalline)