Merck
  • Size effects on the hydrogen storage properties of nanoscaffolded Li3BN2H8.
Size effects on the hydrogen storage properties of nanoscaffolded Li3BN2H8.
Nanotechnology (2009-05-08)
Hui Wu, Wei Zhou, Ke Wang, Terrence J Udovic, John J Rush, Taner Yildirim, Leonid A Bendersky, Adam F Gross, Sky L Van Atta, John J Vajo, Frederick E Pinkerton, Martin S Meyer
ABSTRACT
The use of Li3BN2H8 complex hydride as a practical hydrogen storage material is limited by its high desorption temperature and poor reversibility. While certain catalysts have been shown to decrease the dehydrogenation temperature, no significant improvement in reversibility has been reported thus far. In this study, we demonstrated that tuning the particle size to the nanometer scale by infiltration into nanoporous carbon scaffolds leads to dramatic improvements in the reversibility of Li3BN2H8. Possible changes in the dehydrogenation path were also observed in the nanoscaffolded hydride.
Sigma-Aldrich
Lithium borohydride, hydrogen-storage grade, ≥90%
Sigma-Aldrich
Lithium borohydride, ≥95.0%
Sigma-Aldrich
Lithium borohydride, ≥90%
Sigma-Aldrich
Lithium borohydride solution, 0.5 M in diethyl ether
Sigma-Aldrich
Lithium borohydride solution, 2.0 M in THF