Merck
  • Expression, purification, crystallization and preliminary diffraction analysis of CapF, a capsular polysaccharide-synthesis enzyme from Staphylococcus aureus.
Expression, purification, crystallization and preliminary diffraction analysis of CapF, a capsular polysaccharide-synthesis enzyme from Staphylococcus aureus.
Acta crystallographica. Section F, Structural biology and crystallization communications (2008-06-10)
Takamitsu Miyafusa, Yoshikazu Tanaka, Makoto Kuroda, Toshiko Ohta, Kouhei Tsumoto
ABSTRACT
Capsular polysaccharides (CPs) are important virulence factors of Staphylococcus aureus. The biosynthesis of type 5 and type 8 CPs (CP5 and CP8), which are produced by most clinical isolates of S. aureus, is catalyzed by 16 CP-assembling proteins. One of these proteins is the enzyme CapF, which catalyzes the synthesis of UDP-N-acetyl-L-fucosamine, a component of both CP5 and CP8. Here, the cloning, expression, purification, crystallization and diffraction analysis of CapF are reported. Optimization of the crystallization conditions by differential scanning calorimetry afforded a crystal of selenomethionine-substituted CapF that diffracted to a resolution of 2.80 A. The crystal belongs to space group P3(2)21, with unit-cell parameters a = b = 119.6, c = 129.5 A.
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
SAFC
MES sodium salt
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra