Merck
  • Effect of chromium compounds on sporulation of Eimeria piriformis oocysts.
Effect of chromium compounds on sporulation of Eimeria piriformis oocysts.
Experimental animals (2008-02-08)
Ming-Hsien Li, Hong-Kean Ooi
ABSTRACT
Freshly defecated unsporulated oocysts of Eimeria piriformis from rabbit were treated with various concentrations (1%, 2.5%, 5%, and 10%) of chromium compounds, potassium dichromate, potassium chromate, chromium oxide and chromium nitrate, to examine their effect on sporulation. The sporulation time of oocysts treated with 1 to 10% K(2)Cr(2)O(7) was 28 h. However, much longer sporulation times of about 60 h were required for oocysts treated with 2.5% CrO(3) and Cr(NO(3))(3). Moreover, for oocysts treated with distilled water, 1% K(2)CrO(4) and 10% K(2)CrO(4), the sporulation times required were 216, 156 and 96 h, respectively. Thus, potassium dichromate was found to have higher catalytic activity for the sporulation of E. piriformis oocysts than other chromium compounds.
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, 99%
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, ≥99.99% trace metals basis