Merck
  • The LETM1/YOL027 gene family encodes a factor of the mitochondrial K+ homeostasis with a potential role in the Wolf-Hirschhorn syndrome.
The LETM1/YOL027 gene family encodes a factor of the mitochondrial K+ homeostasis with a potential role in the Wolf-Hirschhorn syndrome.
The Journal of biological chemistry (2004-05-13)
Karin Nowikovsky, Elisabeth M Froschauer, Gabor Zsurka, Jozef Samaj, Siegfried Reipert, Martin Kolisek, Gerlinde Wiesenberger, Rudolf J Schweyen
ABSTRACT
The yeast open reading frames YOL027 and YPR125 and their orthologs in various eukaryotes encode proteins with a single predicted trans-membrane domain ranging in molecular mass from 45 to 85 kDa. Hemizygous deletion of their human homolog LETM1 is likely to contribute to the Wolf-Hirschhorn syndrome phenotype. We show here that in yeast and human cells, these genes encode integral proteins of the inner mitochondrial membrane. Deletion of the yeast YOL027 gene (yol027Delta mutation) results in mitochondrial dysfunction. This mutant phenotype is complemented by the expression of the human LETM1 gene in yeast, indicating a functional conservation of LetM1/Yol027 proteins from yeast to man. Mutant yol027Delta mitochondria have increased cation contents, particularly K+ and low-membrane-potential Deltapsi. They are massively swollen in situ and refractory to potassium acetate-induced swelling in vitro, which is indicative of a defect in K+/H+ exchange activity. Thus, YOL027/LETM1 are the first genes shown to encode factors involved in both K+ homeostasis and organelle volume control.
Sigma-Aldrich
Potassium acetate solution, BioUltra, Molecular Biology, 5 M in H2O
Sigma-Aldrich
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
Sigma-Aldrich
Potassium acetate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%