Merck
  • Bactericidal synergy of lysostaphin in combination with antimicrobial peptides.
Bactericidal synergy of lysostaphin in combination with antimicrobial peptides.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (2011-02-12)
A P Desbois, P J Coote
ABSTRACT
Drug-resistant staphylococci constitute a serious problem that urgently requires the discovery of new therapeutic agents. There has been a resurgence in interest in using lysostaphin (a specific anti-staphylococcal enzyme) as a treatment for infections caused by these important pathogens. However, bacterial resistance to lysostaphin is a problem, but the use of a combination treatment may surmount this issue. In this present study, using viable counts from suspension incubations, lysostaphin is shown to be synergistically bactericidal in combination with various conventional antimicrobial peptides, the antimicrobial protein bovine lactoferrin, a lantibiotic (nisin), and certain lipopeptides used clinically (colistin, daptomycin and polymyxin B). Combinations that act in synergy are of clinical importance as these reduce the doses of the compounds needed for effective treatments and decrease the chances of resistance being selected. The use of lysostaphin in combination with a peptide may represent a new avenue in tackling drug-resistant staphylococci.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein