Merck
PSA forms complexes with α1-antichymotrypsin in prostate.
The Prostate (2012-07-19)
Lei Zhu, Sari Jäämaa, Taija M Af Hällström, Marikki Laiho, Anna Sankila, Stig Nordling, Ulf-Håkan Stenman, Hannu Koistinen
ABSTRACT
PSA is the most useful prostate cancer marker. However, its levels are increased also in some non-malignant conditions. In circulation, the majority of PSA is complexed with protease inhibitors, including α(1) -antichymotrypsin (ACT). The proportion of the PSA-ACT complex is higher in patients with prostate cancer than in controls without cancer. The expression of ACT has been shown to be higher in prostate cancer than in benign prostatic hyperplasia. However, results regarding the extent which PSA forms complexes within the prostate and whether there are differences in complex formation between normal and malignant prostatic tissue are inconsistent and limited. We studied complex formation of PSA secreted by cultured human prostate tissues and in the tissue by in situ proximity ligation assay (PLA). Free, total and active PSA, and the PSA-ACT complex were determined in tissue culture media by immunoassays, immunoblotting, and chromatographic methods. The majority of PSA in tissue culture medium was free and enzymatically active. However, a significant proportion (1.6 ± 0.5%) of immunoreactive PSA was found to be complexed with ACT. Complex formation was confirmed by in situ PLA, which showed more intense staining of PSA-ACT in cancers with Gleason grade 3 than in adjacent benign tissues from the same patients. These results show that PSA forms complexes already within the prostate and that PSA-ACT levels are increased in moderately differentiated prostate cancer tissue. This may explain, at least partially, why the ratio of serum PSA-ACT to total PSA is increased in prostate cancer.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield