Merck
  • Retinoid and thiazolidinedione therapies in melanoma: an analysis of differential response based on nuclear hormone receptor expression.
Retinoid and thiazolidinedione therapies in melanoma: an analysis of differential response based on nuclear hormone receptor expression.
Molecular cancer (2009-03-10)
Joshua P Klopper, Vibha Sharma, Andrew Berenz, William R Hays, Michele Loi, Umarani Pugazhenthi, Sherif Said, Bryan R Haugen
ABSTRACT
Metastatic melanoma has a high mortality rate and suboptimal therapeutic options. Molecular targeting may be beneficial using the rexinoid LGD1069, a retinoid x receptor selective agonist, and thiazolidinediones (TZD), PPARgamma selective ligands, as novel treatments. Mouse xenograft models with human melanoma cell lines [A375(DRO) or M14(5-16)] were treated for 4 weeks with daily vehicle, RXR agonist (rexinoid, LGD1069, 30 mg/kg/d), PPARgamma agonist (TZD, rosiglitazone, 10 mg/kg/d) or combination. A375(DRO) tumor growth was significantly inhibited by either ligand alone and the combination had an additive effect. M14(5-16) tumors only responded to LGD1069 100 mg/kg/day. A375(DRO) sublines resistant to rexinoid, TZD and combination were generated and all three sublines had reduced PPARgamma expression but preserved RXR expression. shRNA knockdown of PPARgamma or RXRgamma attenuated the rexinoid, TZD and combination ligand-mediated decreased proliferation in A375(DRO) cells. Rexinoid (LGD1069) and retinoid (TTNPB) treatment of M14(5-16) cells resulted in decreased proliferation that was additive with combination of both rexinoid and retinoid. shRNA knockdown of RXRgamma resulted in a decreased response to either ligand. A375 (DRO) melanoma cell growth is inhibited by rexinoid and TZD treatment, and this response is dependent on RXR and PPARgamma receptor expression. M14 (5-16) melanoma cell growth is inhibited by rexinoid and retinoid treatment, and this response is dependent on RXR expression. These findings may help guide molecular-based treatment strategies in melanoma and provide insight for mechanisms of resistance to nuclear receptor targeted therapies in certain cancers.
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Lentiviral Particles, Kinases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Lentiviral Particles, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Lentiviral Particles, Tyrosine Kinases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Cytokines and Chemokines
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Tumor Suppressors
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Ubiquitin Hydrolases (DUBS)
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Nuclear Hormone Receptors
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Cytoskeleton Genes
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, p53 Pathway
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Cytokine and Chemokine Receptors
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Extracellular Matrix Genes
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Apoptosis Pathway
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Helicases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Phosphatases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, JAK-STAT Pathway
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, T-Cell Activation
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, B-Cell Activation
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Ion Channels
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, G-Protein Coupled Receptors
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Ubiquitin Ligases (E1,E2,E3)
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Epigenetic Regulators
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, DNA Repair Pathway
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Cell Adhesion Genes
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Kinases
Sigma-Aldrich
TTNPB